Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-(4-methylbenzenesulfonamido)benzoate ≥98%

Supplier: AMBEED, INC
Warning

A866132-10MG A866132-1MG A866132-25MG A866132-50MG A866132-5MG
76654-630EA 11 USD InStock
76654-630 76654-632 76654-634 76654-636 76654-638
Methyl 3-(4-methylbenzenesulfonamido)benzoate ≥98%
Methyl 3-(4-methylbenzenesulfonamido)benzoate
Precaución: for Research Use Only


Formula: C₁₅H₁₅NO₄S
MDL Number: MFCD00994918
CAS Number: 173436-66-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty