Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromo-4-methoxypyrazolo[1,5-a]pyridine-3-carbonitrile 95%

Supplier: AMBEED, INC
Warning

A866632-10G A866632-5G A866632-1G
76654-652EA 53 USD InStock
76654-652 77749-676 77749-675
6-Bromo-4-methoxypyrazolo[1,5-a]pyridine-3-carbonitrile 95%
6-Bromo-4-methoxypyrazolo[1,5-a]pyridine-3-carbonitrile
Precaución: for Research Use Only


Formula: C₉H₆BrN₃O
MDL Number: MFCD27937916
CAS Number: 1207836-10-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty