Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Trifluoromethyl)piperidin-2-one 95%

Supplier: AMBEED, INC
Warning

A868641-100MG A868641-1G A868641-250MG
76654-680EA 10 USD InStock
76654-680 76654-682 76654-684
3-(Trifluoromethyl)piperidin-2-one 95%
3-(Trifluoromethyl)piperidin-2-one
Precaución: for Research Use Only


Formula: C₆H₈F₃NO
MDL Number: MFCD09802218
CAS Number: 1000934-03-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty