Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Thia-6-azaspiro[3.4]octane 2,2-dioxide hydrochloride 98%

Supplier: AMBEED, INC
Warning

A869121-100MG A869121-1G A869121-250MG
76654-692EA 73 USD InStock
76654-692 76654-694 76654-696
2-Thia-6-azaspiro[3.4]octane 2,2-dioxide hydrochloride 98%
2-Thia-6-azaspiro[3.4]octane 2,2-dioxide hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₂ClNO₂S
MDL Number: MFCD30730563
CAS Number: 2095411-01-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty