Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

8-Amino-3,6-dioxaoctanoic acid tert-butyl ester 97%

Supplier: AMBEED, INC
Danger

A879554-250MG A879554-1G A879554-5G
76654-826EA 60 USD InStock
76654-826 77623-098 77623-100
8-Amino-3,6-dioxaoctanoic acid tert-butyl ester 97%
8-Amino-3,6-dioxaoctanoic acid tert-butyl ester
Precaución: for Research Use Only


Formula: C₁₀H₂₁NO₄
MDL Number: MFCD24395885
CAS Number: 1122484-77-8
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty