Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-((3,4-Dichlorobenzyl)amino)propyl)(diethoxymethyl)phosphinic acid 97%

Supplier: AMBEED, INC
Warning

A885418-10MG A885418-1MG A885418-25MG A885418-50MG A885418-5MG
76654-868EA 47 USD InStock
76654-868 76654-870 76654-872 76654-874 76654-876
(3-((3,4-Dichlorobenzyl)amino)propyl)(diethoxymethyl)phosphinic acid 97%
(3-((3,4-Dichlorobenzyl)amino)propyl)(diethoxymethyl)phosphinic acid
Precaución: for Research Use Only


Formula: C₁₅H₂₄Cl₂NO₄P
MDL Number: MFCD09878290
CAS Number: 139667-74-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty