Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(((2R,3R,4R,5R,6R)-3-Acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)pentanoic acid 97%

Supplier: AMBEED, INC
Warning

A885647-100MG A885647-50MG
76654-884EA 10 USD InStock
76654-884 76654-886
5-(((2R,3R,4R,5R,6R)-3-Acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)pentanoic acid 97%
5-(((2R,3R,4R,5R,6R)-3-Acetamido-4,5-diacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)pentanoic acid
Precaución: for Research Use Only


Formula: C₁₉H₂₉NO₁₁
MDL Number: MFCD30802159
CAS Number: 1159408-54-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty