Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Chloromethyl)-2-fluorobenzonitrile 95%

Supplier: AMBEED, INC
Danger

A886178-1G A886178-250MG
76654-898EA 87 USD InStock
76654-898 76654-900
5-(Chloromethyl)-2-fluorobenzonitrile 95%
5-(Chloromethyl)-2-fluorobenzonitrile
Precaución: for Research Use Only


Formula: C₈H₅ClFN
MDL Number: MFCD12756602
CAS Number: 1260892-59-8
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty