Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium Pyridine-4-sulfinate 97%

Supplier: AMBEED, INC
Warning

A886251-100MG A886251-1G A886251-250MG
76654-902EA 89 USD InStock
76654-902 76654-904 76654-906
Sodium Pyridine-4-sulfinate 97%
Sodium Pyridine-4-sulfinate
Precaución: for Research Use Only


Formula: C₅H₄NNaO₂S
MDL Number: MFCD30502924
CAS Number: 116008-37-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty