Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyromellitic acid 98%

Supplier: AMBEED, INC
Warning

A887143-1000G A887143-100G A887143-25G A887143-500G
76654-926EA 29 USD InStock
76654-926 76654-928 76654-930 76654-932
Pyromellitic acid 98%
Pyromellitic acid
Precaución: for Research Use Only


Formula: C₁₀H₆O₈
MW: 254.15 g/mol
Boiling Pt: 516
Melting Pt: 272
MDL Number: MFCD00002471
CAS Number: 89-05-4
Merck Index: 12,08188
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty