Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-carboxymethyl-3-methylimidazolium chloride 99%

Supplier: AMBEED, INC
Warning

A895785-100G A895785-25G A895785-5G
76655-052EA 82 USD InStock
76655-052 76655-054 76655-056 76655-058
1-carboxymethyl-3-methylimidazolium chloride 99%
1-carboxymethyl-3-methylimidazolium chloride
Precaución: for Research Use Only


Formula: C₆H₉ClN₂O₂
MDL Number: MFCD27963380
CAS Number: 700370-07-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty