Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

tert-Butyl 18-bromooctadecanoate 97%

Supplier: AMBEED, INC
Warning

A899898-100MG A899898-250MG A899898-25G
76655-098EA 19 USD InStock
76655-098 76655-100 77749-707
tert-Butyl 18-bromooctadecanoate 97%
tert-Butyl 18-bromooctadecanoate
Precaución: for Research Use Only


Formula: C₂₂H₄₃BrO₂
MW: 419.4796 g/mol
MDL Number: MFCD31727993
CAS Number: 1199580-32-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty