Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-Acetylphenyl)-2-(4-(2,6-dioxo-1,3-dipropyl-2,3,6,9-tetrahydro-1H-purin-8-yl)phenoxy)acetamide 98%

Supplier: AMBEED, INC
Warning

A900481-10MG A900481-1MG A900481-50MG A900481-5MG
76655-114EA 160 USD InStock
76655-114 76655-116 76655-118 76655-120
N-(4-Acetylphenyl)-2-(4-(2,6-dioxo-1,3-dipropyl-2,3,6,9-tetrahydro-1H-purin-8-yl)phenoxy)acetamide 98%
N-(4-Acetylphenyl)-2-(4-(2,6-dioxo-1,3-dipropyl-2,3,6,9-tetrahydro-1H-purin-8-yl)phenoxy)acetamide
Precaución: for Research Use Only


Formula: C₂₇H₂₉N₅O₅
MDL Number: MFCD06798374
CAS Number: 264622-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty