Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-5069 98%

Supplier: AMBEED, INC
Danger

A912767-10MG A912767-1MG A912767-25MG A912767-5MG
76655-244EA 40 USD InStock
76655-244 76655-246 76655-248 76655-250
AZD-5069 98%
AZD-5069
Precaución: for Research Use Only


Formula: C₁₈H₂₂F₂N₄O₅S₂
MDL Number: MFCD19443645
CAS Number: 878385-84-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty