Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-(5,6-Difluoro-2-methyl-1H-benzo[d]imidazol-1-yl)-N-(4-(trifluoromethyl)phenyl)pyrazin-2-amine 98%

Supplier: AMBEED, INC
Warning

A914586-1MG A914586-25MG A914586-5MG
76655-272EA 20 USD InStock
76655-272 76655-274 76655-276
6-(5,6-Difluoro-2-methyl-1H-benzo[d]imidazol-1-yl)-N-(4-(trifluoromethyl)phenyl)pyrazin-2-amine 98%
6-(5,6-Difluoro-2-methyl-1H-benzo[d]imidazol-1-yl)-N-(4-(trifluoromethyl)phenyl)pyrazin-2-amine
Precaución: for Research Use Only


Formula: C₁₉H₁₂F₅N₅
MDL Number: MFCD28716152
CAS Number: 1782970-28-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty