Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-N-(4H-1,2,4-triazol-4-yl)benzamide 90%

Supplier: AMBEED, INC
Warning

A916019-100MG A916019-1G A916019-250MG
76655-296EA 26 USD InStock
76655-296 76655-298 76655-300
4-Fluoro-N-(4H-1,2,4-triazol-4-yl)benzamide 90%
4-Fluoro-N-(4H-1,2,4-triazol-4-yl)benzamide
Precaución: for Research Use Only


Formula: C₉H₇FN₄O
MDL Number: MFCD00448017
CAS Number: 331434-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty