Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(8-Chloroquinolin-6-yl)boronic acid 98%

Supplier: AMBEED, INC
Danger

A917960-100MG A917960-1G A917960-250MG
76655-352EA 37 USD InStock
76655-352 76655-354 76655-356
(8-Chloroquinolin-6-yl)boronic acid 98%
(8-Chloroquinolin-6-yl)boronic acid
Precaución: for Research Use Only


Formula: C₉H₇BClNO₂
MDL Number: MFCD27935582
CAS Number: 2230902-55-1
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty