Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-2-Dodecen-1-ol 95% (by GC)

Supplier: AMBEED, INC
Warning

A917989-1G A917989-250MG
76655-358EA 54 USD InStock
76655-358 77623-138
trans-2-Dodecen-1-ol 95% (by GC)
trans-2-Dodecen-1-ol
Precaución: for Research Use Only


Formula: C₁₂H₂₄O
MDL Number: MFCD00014047
CAS Number: 69064-37-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty