Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Guanylurea phosphate 98% (by titrimetric analysis)

Supplier: AMBEED, INC
Warning

A921312-100G A921312-25G A921312-500G
76655-398EA 12 USD InStock
76655-398 76655-400 76655-402
Guanylurea phosphate 98% (by titrimetric analysis)
Guanylurea phosphate
Precaución: for Research Use Only


Formula: C₂H₆N₄O·H₃PO₄
MDL Number: MFCD00065097
CAS Number: 36897-89-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty