Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Ethanedisulfonic acid dihydrate 97%

Supplier: AMBEED, INC
Danger

A921370-1G A921370-25G A921370-5G
76655-408EA 141.1 USD InStock
76655-408 76655-410 76655-412 76655-414
1,2-Ethanedisulfonic acid dihydrate 97%
1,2-Ethanedisulphonic acid dihydrate
Precaución: for Research Use Only


Formula: C₂H₆O₆S₂·2H₂O
MDL Number: MFCD26793415
CAS Number: 5982-56-9
UN: 2585
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty