Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Remetinostat 99%

Supplier: AMBEED, INC
Danger

A922480-10MG A922480-1MG A922480-25MG A922480-50MG A922480-5MG
76655-422EA 55 USD InStock
76655-422 76655-424 76655-426 76655-428 76655-430
Remetinostat 99%
Remetinostat
Precaución: for Research Use Only


Formula: C₁₆H₂₁NO₆
MW: 323.34 g/mol
MDL Number: MFCD28986227
CAS Number: 946150-57-8
UN: 1325
ADR: 4.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty