Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-(bromomethyl)-5-fluoro-3-nitrobenzoate 98%

Supplier: AMBEED, INC
Danger

A922481-100MG A922481-1G A922481-250MG
76655-432EA 54 USD InStock
76655-432 76655-434 76655-436
Methyl 2-(bromomethyl)-5-fluoro-3-nitrobenzoate 98%
Methyl 2-(bromomethyl)-5-fluoro-3-nitrobenzoate
Precaución: for Research Use Only


Formula: C₉H₇BrFNO₄
MW: 292.0586 g/mol
MDL Number: MFCD25977284
CAS Number: 850462-65-6
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty