Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(3-Chloro-4-methanesulfonylphenyl)boronic acid 96%

Supplier: AMBEED, INC
Warning

A924469-100MG A924469-1G A924469-250MG
76655-496EA 37 USD InStock
76655-496 76655-498 76655-500
(3-Chloro-4-methanesulfonylphenyl)boronic acid 96%
(3-Chloro-4-methanesulfonylphenyl)boronic acid
Precaución: for Research Use Only


Formula: C₇H₈BClO₄S
MDL Number: MFCD27935715
CAS Number: 2239305-86-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty