Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Iodostyrene 97% stabilized

Supplier: AMBEED, INC
Warning

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A926828-1G A926828-250MG A926828-5G A926828-100MG
76655-514EA 165 USD InStock
76655-514 76655-516 76655-518 77595-496
4-Iodostyrene 97% stabilized
4-Iodostyrene
Precaución: for Research Use Only


Formula: C₈H₇I
MDL Number: MFCD00045321
CAS Number: 2351-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty