Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(Benzyloxy)-1-(2,2-dimethoxyethyl)-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A928015-10G A928015-1G A928015-250MG A928015-25G A928015-5G
76655-528EA 263 USD InStock
76655-528 76655-530 76655-532 76655-534 76655-536
5-(Benzyloxy)-1-(2,2-dimethoxyethyl)-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid 97%
5-(Benzyloxy)-1-(2,2-dimethoxyethyl)-6-(methoxycarbonyl)-4-oxo-1,4-dihydropyridine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₉H₂₁NO₈
CAS Number: 1973401-99-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty