Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethoxy)phenylmethanesulfonyl chloride 95%

Supplier: AMBEED, INC
Danger

A930482-250MG A930482-1G
76655-570EA 47 USD InStock
76655-570 77623-162
4-(Trifluoromethoxy)phenylmethanesulfonyl chloride 95%
4-(Trifluoromethoxy)phenylmethanesulfonyl chloride
Precaución: for Research Use Only


Formula: C₈H₆ClF₃O₃S
MDL Number: MFCD09811274
CAS Number: 683813-55-0
UN: 3261
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty