Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-((2-Chlorophenyl)thio)-4-hydroxy-6-(4-morpholinophenyl)-6-(thiophen-3-yl)-5,6-dihydropyridin-2(1H)-one 98%

Supplier: AMBEED, INC
Warning

A932685-10MG A932685-1MG A932685-50MG A932685-5MG A932685-100MG
76655-582EA 298 USD InStock
76655-582 76655-584 76655-586 76655-588 77623-164
(R)-3-((2-Chlorophenyl)thio)-4-hydroxy-6-(4-morpholinophenyl)-6-(thiophen-3-yl)-5,6-dihydropyridin-2(1H)-one 98%
(R)-3-((2-Chlorophenyl)thio)-4-hydroxy-6-(4-morpholinophenyl)-6-(thiophen-3-yl)-5,6-dihydropyridin-2(1H)-one
Precaución: for Research Use Only


Formula: C₂₅H₂₃ClN₂O₃S₂
MDL Number: MFCD31560470
CAS Number: 2003234-63-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty