Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Propargylmaleimide 98% (by HPLC, total nitrogen)

Supplier: AMBEED, INC
Warning

A932844-100MG A932844-1G A932844-250MG A932844-5G
76655-590EA 19 USD InStock
76655-590 76655-592 76655-594 77623-166
N-Propargylmaleimide 98% (by HPLC, total nitrogen)
N-Propargylmaleimide
Precaución: for Research Use Only


Formula: C₇H₅NO₂
MDL Number: MFCD12090919
CAS Number: 209395-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty