Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dimethylisophthalic acid 98%

Supplier: AMBEED, INC
Warning

A936930-100MG A936930-1G A936930-250MG A936930-5G
76655-686EA 53 USD InStock
76655-686 76655-688 76655-690 76655-692
4,6-Dimethylisophthalic acid 98%
4,6-Dimethylisophthalic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₄
MDL Number: MFCD00230851
CAS Number: 2790-09-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty