Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Diacetylbenzene 95%

Supplier: AMBEED, INC
Warning

A943264-1G A943264-250MG A943264-25G A943264-5G
76655-860EA 15 USD InStock
76655-860 76655-862 76655-864 76655-866
1,4-Diacetylbenzene 95%
1,4-Diacetylbenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₀O₂
MDL Number: MFCD00008750
CAS Number: 1009-61-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty