Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine 90%

Supplier: AMBEED, INC
Warning

A943764-100MG A943764-1G A943764-250MG
76655-886EA 35 USD InStock
76655-886 76655-888 76655-890
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine 90%
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]piperazine
Precaución: for Research Use Only


Formula: C₉H₁₅N₅O₂
MDL Number: MFCD16076302
CAS Number: 1248142-10-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty