Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-tert-Butyl 2-methylaziridine-1-carboxylate 97%

Supplier: AMBEED, INC
Danger

A943853-100MG A943853-1G A943853-250MG
76655-892EA 12 USD InStock
76655-892 76655-894 76655-896
(R)-tert-Butyl 2-methylaziridine-1-carboxylate 97%
(R)-tert-Butyl 2-methylaziridine-1-carboxylate
Precaución: for Research Use Only


Formula: C₈H₁₅NO₂
MW: 157.2102 g/mol
MDL Number: MFCD28098437
CAS Number: 129319-91-1
UN: 1993
ADR: 3 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty