Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Erucic acid 85%

Supplier: AMBEED, INC
Warning

A949077-100G A949077-25G A949077-500G A949077-1000G
76655-964EA 41 USD InStock
76655-964 76655-966 76655-968 76655-970 77623-174
Erucic acid 85%
Erucic acid
Precaución: for Research Use Only


Formula: C₂₂H₄₂O₂
MDL Number: MFCD00063188
CAS Number: 112-86-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty