Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[2,3-c]pyridin-7-ylmethanol 97%

Supplier: AMBEED, INC
Warning

A950631-100MG A950631-1G A950631-250MG
76656-018EA 119 USD InStock
76656-018 76656-020 76656-022
1H-Pyrrolo[2,3-c]pyridin-7-ylmethanol 97%
1H-Pyrrolo[2,3-c]pyridin-7-ylmethanol
Precaución: for Research Use Only


Formula: C₈H₈N₂O
MDL Number: MFCD22373669
CAS Number: 1379526-98-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty