Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5,6,7-Tetrahydro-1H-pyrazolo[4,3-b]pyridine hydrochloride 96%

Supplier: AMBEED, INC
Warning

A954598-100MG A954598-250MG
76656-074EA 23 USD InStock
76656-074 76656-076
4,5,6,7-Tetrahydro-1H-pyrazolo[4,3-b]pyridine hydrochloride 96%
4,5,6,7-Tetrahydro-1H-pyrazolo[4,3-b]pyridine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₀ClN₃
MDL Number: MFCD26792521
CAS Number: 2156657-30-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty