Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Tetrahydro-2H-pyran-4-yl)methoxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A955043-100MG A955043-1G A955043-250MG
76656-082EA 39 USD InStock
76656-082 76656-084 76656-086
4-(Tetrahydro-2H-pyran-4-yl)methoxyphenylboronic acid 98%
4-(Tetrahydro-2H-pyran-4-yl)methoxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₇BO₄
MDL Number: MFCD17474507
CAS Number: 1615247-95-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty