Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole 98% (by HPLC)

Supplier: AMBEED, INC
Warning

A958031-1G A958031-250MG A958031-5G
76656-136EA 51 USD InStock
76656-136 76656-138 76656-140
9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole 98% (by HPLC)
9,9'-Diphenyl-9H,9'H-3,3'-bicarbazole
Precaución: for Research Use Only


Formula: C₃₆H₂₄N₂
MDL Number: MFCD29905037
CAS Number: 57102-62-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty