Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triethylamine phosphate 96% (by titrimetric analysis)

Supplier: AMBEED, INC
Warning

A962688-100G A962688-10G A962688-1G A962688-25G A962688-500G A962688-5G
76656-184EA 194 USD InStock
76656-184 76656-186 76656-188 76656-190 76656-192 76656-194
Triethylamine phosphate 96% (by titrimetric analysis)
Triethylamine phosphate
Precaución: for Research Use Only


Formula: C₆H₁₅N
MW: 199.19 g/mol
MDL Number: MFCD00067475
CAS Number: 35365-94-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty