Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(±)-Evodiamine 98%

Supplier: AMBEED, INC
Warning

A964394-1G A964394-25G A964394-5G
76656-234EA 18 USD InStock
76656-234 77623-202 77623-204
(±)-Evodiamine 98%
(±)-Evodiamine
Precaución: for Research Use Only


Formula: C₁₉H₁₇N₃O
MDL Number: MFCD06407824
CAS Number: 518-18-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty