Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxyethyl 2-bromoisobutyrate 97%

Supplier: AMBEED, INC
Danger

A965100-100MG A965100-1G A965100-250MG A965100-5G
76656-250EA 24 USD InStock
76656-250 76656-252 76656-254 76656-256
2-Hydroxyethyl 2-bromoisobutyrate 97%
2-Hydroxyethyl 2-bromoisobutyrate
Precaución: for Research Use Only


Formula: C₆H₁₁BrO₃
MW: 211.0537 g/mol
MDL Number: MFCD16621490
CAS Number: 189324-13-8
UN: 3265
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty