Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-1,1-difluorocyclohexane 97%

Supplier: AMBEED, INC
Warning

A966129-100MG A966129-250MG A966129-5G
76656-302EA 68 USD InStock
76656-302 76656-304 76656-306
4-Bromo-1,1-difluorocyclohexane 97%
4-Bromo-1,1-difluorocyclohexane
Precaución: for Research Use Only


Formula: C₆H₉BrF₂
MDL Number: MFCD13189572
CAS Number: 1196156-51-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty