Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Strontium sulphate 98%

Supplier: AMBEED, INC
Warning

A967158-1000G A967158-100G A967158-25G A967158-500G
76656-328EA 34 USD InStock
76656-328 76656-330 76656-332 76656-334
Strontium sulphate 98%
Strontium sulphate
Precaución: for Research Use Only


Formula: O₄SSr
MW: 183.68
Melting Pt: 1600
Density: 3.96
MDL Number: MFCD00036274
CAS Number: 7759-02-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty