Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dimethyl-2-(trimethylsilyl)phenyl trifluoromethanesulfonate 97%

Supplier: AMBEED, INC
Danger

A968337-100MG A968337-250MG
76656-338EA 43 USD InStock
76656-338 76656-340
4,5-Dimethyl-2-(trimethylsilyl)phenyl trifluoromethanesulfonate 97%
4,5-Dimethyl-2-(trimethylsilyl)phenyl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₁₂H₁₇F₃O₃SSi
MDL Number: MFCD28969391
CAS Number: 458566-99-9
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty