Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

22-(tert-Butoxy)-22-oxodocosanoic acid 97%

Supplier: AMBEED, INC
Warning

A971470-1G A971470-250MG
76656-404EA 31 USD InStock
76656-404 76656-406
22-(tert-Butoxy)-22-oxodocosanoic acid 97%
22-(tert-Butoxy)-22-oxodocosanoic acid
Precaución: for Research Use Only


Formula: C₂₆H₅₀O₄
MDL Number: MFCD30830215
CAS Number: 1642333-05-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty