Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,5,5-Tetramethyl-2-(2,4,6-trifluorophenyl)-1,3,2-dioxaborolane 97%

Supplier: AMBEED, INC
Warning

A975013-250MG A975013-5G
76656-434EA 11 USD InStock
76656-434 76656-436
4,4,5,5-Tetramethyl-2-(2,4,6-trifluorophenyl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(2,4,6-trifluorophenyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₂H₁₄BF₃O₂
MDL Number: MFCD12405351
CAS Number: 325143-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty