Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5-Tris[4-(9-carbazolyl)phenyl]benzene 98% (by HPLC, total nitrogen)

Supplier: AMBEED, INC
Warning

A976276-100MG A976276-1G A976276-250MG
76656-454EA 28 USD InStock
76656-454 76656-456 76656-458
1,3,5-Tris[4-(9-carbazolyl)phenyl]benzene 98% (by HPLC, total nitrogen)
1,3,5-Tris[4-(9-carbazolyl)phenyl]benzene
Precaución: for Research Use Only


Formula: C₆₀H₃₉N₃
MDL Number: MFCD29765751
CAS Number: 160780-82-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty