Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Chloropyridin-3-yl)-2-methylpropanoic acid 95%

Supplier: AMBEED, INC
Danger

A979557-250MG A979557-5G
76656-504EA 102 USD InStock
76656-504 76656-506
2-(2-Chloropyridin-3-yl)-2-methylpropanoic acid 95%
2-(2-Chloropyridin-3-yl)-2-methylpropanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀ClNO₂
MDL Number: MFCD31744070
CAS Number: 1452561-74-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty