Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-Iodopyrrolo[2,1-f][1,2,4]triazin-4-amine 97%

Supplier: AMBEED, INC
Warning

A979968-100G A979968-1G A979968-25G A979968-5G
76656-526EA 247 USD InStock
76656-526 76656-528 76656-530 76656-532
7-Iodopyrrolo[2,1-f][1,2,4]triazin-4-amine 97%
7-Iodopyrrolo[2,1-f][1,2,4]triazin-4-amine
Precaución: for Research Use Only


Formula: C₆H₅IN₄
MDL Number: MFCD21607515
CAS Number: 1770840-43-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty