Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (3E)-1-acetyl-3-[methoxy(phenyl)methylidene]-2-oxo-2,3-dihydro-1H-indole-6-carboxylate 96%

Supplier: AMBEED, INC
Warning

A980116-100MG A980116-1G A980116-250MG
76656-540EA 22 USD InStock
76656-540 76656-542 76656-544
Methyl (3E)-1-acetyl-3-[methoxy(phenyl)methylidene]-2-oxo-2,3-dihydro-1H-indole-6-carboxylate 96%
Methyl (3E)-1-acetyl-3-[methoxy(phenyl)methylidene]-2-oxo-2,3-dihydro-1H-indole-6-carboxylate
Precaución: for Research Use Only


Formula: C₂₀H₁₇NO₅
MDL Number: MFCD18207134
CAS Number: 1168152-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty