Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3,5-dichloro-6-hydroxypicolinate 97%

Supplier: AMBEED, INC
Warning

A981433-100MG A981433-1G A981433-250MG
76656-570EA 30 USD InStock
76656-570 76656-572 76656-574
Methyl 3,5-dichloro-6-hydroxypicolinate 97%
Methyl 3,5-dichloro-6-hydroxypicolinate
Precaución: for Research Use Only


Formula: C₇H₅Cl₂NO₃
MDL Number: MFCD31922976
CAS Number: 2368871-72-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty